Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXTL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572069
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572069 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572069 Supplier Invitrogen™ Supplier No. PA572069
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EXTL1 Polyclonal Antibody for Western Blot

This gene is a member of the multiple exostoses (EXT) family of glycosyltransferases, which function in the chain polymerization of heparan sulfate and heparin. The encoded protein harbors alpha 1,4- N-acetylglucosaminyltransferase activity, and is involved in chain elongation of heparan sulfate and possibly heparin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXTL1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Extl1
Gene Accession No. Q92935, Q9JKV7
Gene Alias alpha-N-acetylglucosaminyltransferase II; D430033M16Rik; EC 2.4.1.224; exostoses (multiple)-like 1; exostoses-like 1; exostosin like glycosyltransferase 1; exostosin-L; Exostosin-like 1; Exostosin-like 1 (Exostosin-L) (Multiple exostosis-like protein); exostosin-like glycosyltransferase 1; EXTL; Extl1; glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N- acetylglucosaminyltransferase; glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; Glucuronosyl-N-acetylglucosaminyl-proteoglycan4-alpha-N-acetylglucosaminyltransferase; glucuronyl-N-acetylglucosaminylproteoglycan alpha-1,4-N- acetylglucosaminyltransferase; MGC70794; multiple exostoses-like 1; multiple exostosis-like protein
Gene Symbols Extl1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 649-676 amino acids from the C-terminal region of human EXTL1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2134, 56219
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.