Learn More
Invitrogen™ EXTL2 Polyclonal Antibody

Description
Immunogen sequence: TANRMRNRLQV FPELETNAVL MVDDDTLIST PDLVFAFSVW QQFPDQIVGF VPRKHVSTSS GIYSYGSFEM Highest antigen sequence identity to the following orthologs - mouse 89%, rat 90%.
Specifications
Specifications
| Antigen | EXTL2 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | EXTL2 |
| Gene Accession No. | Q9UBQ6 |
| Gene Alias | 3000001D04Rik; alpha-1,4-N-acetylhexosaminyltransferase EXTL2; Alpha-GalNAcT EXTL2; AW146439; exostoses (multiple)-like 2; exostoses-like 2; exostosin like glycosyltransferase 2; exostosin-like 2; exostosin-like glycosyltransferase 2; exotoses (multiple)-like 2; EXTL2; Extr2; EXT-related protein 2; Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; processed exostosin-like 2 |
| Gene Symbols | EXTL2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.