Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXTL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530025
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530025 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530025 Supplier Invitrogen™ Supplier No. PA530025
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG, MCF-7. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (95%), Xenopus laevis (96%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

EXTL3 is probable glycosyltransferase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXTL3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene EXTL3
Gene Accession No. O43909
Gene Alias 2900009G18Rik; AU040105; BOTV; exostoses (multiple)-like 3; exostoses-like 3; exostosin; exostosin like glycosyltransferase 3; exostosin tumor-like 3; exostosin-like 3; exostosin-like glycosyltransferase 3; Ext1l; EXTL1L; EXTL3; EXTR1; EXT-related 1; EXT-related protein 1; Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; hereditary multiple exostoses gene isolog; KIAA0519; mKIAA0519; multiple exostosis-like protein 3; Putative tumor suppressor protein EXTL3; REG; reg receptor; REGR; RPR
Gene Symbols EXTL3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human EXTL3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2137
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.