Learn More
Invitrogen™ EXTL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120872
Description
Positive test controls include: 293T, Mouse pancreas, Rat lung. The target is usually found in the following locations: Endoplasmic reticulum membrane, Single-pass type II membrane protein. Immunogen sequence: TTLDEADEAG KRIFGPRVGN ELCEVKHVLD LCRIRESVSE ELLQLEAKRQ ELNSEIAKLN LKIEACKKSI ENAKQDLLQL KNVISQTEHS YKELMAQNQP KLSLPIRLLP EKDDAGLPPP KATRGCRLHN CFDYSRCPLT SGFPVYVYDS DQFVFGSYLD PLVKQAFQA.
EXTL3 is probable glycosyltransferase.
Specifications
EXTL3 | |
Polyclonal | |
Unconjugated | |
EXTL3 | |
2900009G18Rik; AU040105; BOTV; exostoses (multiple)-like 3; exostoses-like 3; exostosin; exostosin like glycosyltransferase 3; exostosin tumor-like 3; exostosin-like 3; exostosin-like glycosyltransferase 3; Ext1l; EXTL1L; EXTL3; EXTR1; EXT-related 1; EXT-related protein 1; Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; hereditary multiple exostoses gene isolog; KIAA0519; mKIAA0519; multiple exostosis-like protein 3; Putative tumor suppressor protein EXTL3; REG; reg receptor; REGR; RPR | |
Rabbit | |
Affinity chromatography | |
RUO | |
2137, 54616, 56819 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43909, Q9WVL6 | |
EXTL3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 52-220 of human EXTL3 (NP_001431.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.