Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ringers Extrication Hybrid Gloves

Catalog No. 19822139 Shop All Ansell Products
Encompass
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-822-139 X-Large
19-822-136 Small
19-822-137 Medium
19-822-138 Large
19-822-140 2X-Large
19-822-141 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-822-139 Supplier Ringers Supplier No. 33711
Add to Cart
Add to Cart

Specifications

Color Green
Cut Resistance Rating ANSI A3
Product Type Extrication Gloves
Size X-Large
For Use With (Application) Firemen, Rescue Workers, Military, and Law Enforcement
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.