Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EYA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117158
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117158 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117158 Supplier Invitrogen™ Supplier No. PA5117158
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: SH-SY5Y, 293T, Mouse testis, Mouse brain, Rat brain Immunogen sequence: NSLTNSSGFN SSQQDYPSYP SFGQGQYAQY YNSSPYPAHY MTSSNTSPTT PSTNATYQLQ EPPSGITSQA VTDPTAEYST IHSPSTPIKD SDSDRLRRGS D.

This gene encodes a member of the eyes absent family of proteins. The encoded protein may play a role in the developing kidney, branchial arches, eye, and ear. Mutations of this gene have been associated with branchiootorenal dysplasia syndrome, branchiootic syndrome, and sporadic cases of congenital cataracts and ocular anterior segment anomalies. A similar protein in mice can act as a transcriptional activator. Four transcript variants encoding three distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EYA1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene EYA1
Gene Accession No. P97767, Q99502
Gene Alias BOP; bor; BOS1; dog eared; EYA transcriptional coactivator and phosphatase 1; eya1; eya1 protein; Eyes absent; eyes absent 1; eyes absent 1 homolog; eyes absent 1 homolog (Drosophila); eyes absent homolog 1; MGC141875; OFC1; OTTHUMP00000195053; wu:fc13c10; zgc:100770
Gene Symbols EYA1
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14048, 2138, 502935
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.