Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EYA2 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP146816IR
Description
EYA2 Polyclonal antibody specifically detects EYA2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
EYA2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
EAB1EC 3.1.3.48, eyes absent (Drosophila) homolog 2, eyes absent homolog 2, eyes absent homolog 2 (Drosophila), MGC10614 | |
The immunogen for this product maps to a region between residue 1 and 50 of human eyes absent homolog 2 using the numbering given in entry NP_005235.3 (GeneID 2139). | |
0.1 mL | |
Protein Phosphatase, Vision | |
2139 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction