Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EYA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566243
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566243 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566243 Supplier Invitrogen™ Supplier No. PA566243
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVELVISPSLT VNSDCLDKLK FNRADAAVWT LSDRQGITKS APLRVSQLFS RSCPRVLPRQ PSTAMAAYGQ TQYSAGIQQA TPYTAYPPPA QAYGIPSYSI Highest antigen sequence identity to the following orthologs - mouse 67%, rat 63%.

Functions both as protein phosphatase and as transcriptional coactivator for SIX1, and probably also for SIX2, SIX4 and SIX5. Tyrosine phosphatase that dephosphorylates 'Tyr- 142' of histone H2AX (H2AXY142ph) and promotes efficient DNA repair via the recruitment of DNA repair complexes containing MDC1. 'Tyr-142' phosphorylation of histone H2AX plays a central role in DNA repair and acts as a mark that distinguishes between apoptotic and repair responses to genotoxic stress. Its function as histone phosphatase may contribute to its function in transcription regulation during organogenesis. Plays an important role in hypaxial muscle development together with SIX1 and DACH2; in this it is functionally redundant with EYA1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EYA2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EYA2
Gene Accession No. O00167
Gene Alias Drosophila-type eyes absent 2-like protein; Eab1; EYA transcriptional coactivator and phosphatase 2; Eya2; eyes absent 2; eyes absent 2 homolog; eyes absent 2 homolog (Drosophila); eyes absent 2 isoform a; eyes absent homolog 2
Gene Symbols EYA2
Host Species Rabbit
Immunogen Recombinant Human EYA2. Recombinant protein control fragment (Product #RP-105124).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2139
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.