Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EYA3 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP146819APC
Description
EYA3 Polyclonal antibody specifically detects EYA3 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)Specifications
EYA3 | |
Polyclonal | |
Immunoprecipitation, Western Blot (Negative) | |
DKFZp686C132, EC 3.1.3.48, eyes absent (Drosophila) homolog 3, eyes absent 3, eyes absent homolog 3, eyes absent homolog 3 (Drosophila) | |
The immunogen for this product maps to a region between residue 30 and 60 of human eyes absent homolog 3 using the numbering given in entry NP_001981.2 (GeneID 2140). | |
0.1 mL | |
Vision | |
2140 | |
Store at 4C in the dark. | |
IgG |
Immunoprecipitation | |
APC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction