Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EYA3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100511
Description
Antibody detects endogenous levels of total EYA3.
Eyes absent homolog 3 (EYA3) is a member of the eyes absent (EYA) family of proteins. This protein may act as a transcriptional activator and have a role during development. A similar protein in mice can act as a transcriptional activator. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
EYA3 | |
Polyclonal | |
Unconjugated | |
Eya3 | |
AI844637; EYA transcriptional coactivator and phosphatase 3; EYA3; eyes absent 3; eyes absent 3 homolog; eyes absent 3 homolog (Drosophila); eyes absent homolog 3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
14050, 2140, 313027 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P97480, Q99504 | |
Eya3 | |
A synthesized peptide derived from human EYA3(Accession Q99504), corresponding to amino acid residues S262-E312. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction