Learn More
EYA3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc IHC00738

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Sheep, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan, Monkey,
Eyes absent homolog 3 (EYA3) is a member of the eyes absent (EYA) family of proteins. This protein may act as a transcriptional activator and have a role during development. A similar protein in mice can act as a transcriptional activator. Two transcript variants encoding distinct isoforms have been identified for this gene.Specifications
EYA3 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q99504 | |
EYA3 | |
Between 1 and 30 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry | |
Unconjugated | |
EYA3 | |
Eyes Absent Homolog 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2140 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.