Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EYA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | EYA3 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3313
![]() ![]() |
Proteintech Group Inc
211961AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is specific to EYA3.
Eyes absent homolog 3 (EYA3) is a member of the eyes absent (EYA) family of proteins. This protein may act as a transcriptional activator and have a role during development. A similar protein in mice can act as a transcriptional activator. Two transcript variants encoding distinct isoforms have been identified for this gene.Specifications
EYA3 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P97480, Q99504 | |
14050, 2140, 313027 | |
EYA3 Fusion Protein Ag15544 | |
Primary | |
-20°C | |
EYA3 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
EYA3, Eyes absent homolog 3 | |
Eya3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title