Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EYA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558534
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558534 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558534 Supplier Invitrogen™ Supplier No. PA558534
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QYAQYYSAST YGAYMTSNNT ADGTPSSTST YQLQESLPGL TNQPGEFDTM QSPSTPIKDL DERTCRSSGS KS Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

This gene encodes a member of the eyes absent (EYA) family of proteins. The encoded protein may act as a transcriptional activator through its protein phosphatase activity, and it may be important for eye development, and for continued function of the mature organ of Corti. Mutations in this gene are associated with postlingual, progressive, autosomal dominant hearing loss at the deafness, autosomal dominant non-syndromic sensorineural 10 locus. The encoded protein is also a putative oncogene that mediates DNA repair, apoptosis, and innate immunity following DNA damage, cellular damage, and viral attack. Defects in this gene are also associated with dilated cardiomyopathy 1J. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EYA4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EYA4
Gene Accession No. O95677, Q9Z191
Gene Alias B130023L16Rik; CMD1J; DFNA10; dJ78N10.1 (eyes absent); EYA transcriptional coactivator and phosphatase 4; EYA4; eyes absent 4 homolog (Drosophila); eyes absent homolog 4; eyes absent-like protein 4
Gene Symbols EYA4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EYA4. Recombinant protein control fragment (Product #RP-94980).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14051, 2070, 292172
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.