Learn More
EYA4 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302628A

Description
The recommended shelf life for this product is 1 year from date of receipt.
EYA4 encodes a member of the eyes absent (EYA) family of proteins. The encoded protein may act as a transcriptional activator through its protein phosphatase activity, and it may be important for eye development, and for continued function of the mature organ of Corti.Specifications
EYA4 | |
Polyclonal | |
Unconjugated | |
EYA4 | |
CMD1J, DFNA10, dJ78N10.1 (eyes absent), eyes absent homolog 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2070 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O95677 | |
EYA4 | |
Between 1 and 50 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.