Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EYS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555507
Description
Immunogen sequence: CECTSGWTGQ NCSEEINECD SDPCMNGGLC HESTIPGQFV CLCPPLYTGQ FCHQRYNLCD LLHNPCR Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 46%.

Specifications
EYS | |
Polyclonal | |
Unconjugated | |
EYS | |
bA166P24.2; bA307F22.3; bA74E24.1; C6orf178; C6orf179; C6orf180; dJ1018A4.2; dJ22I17.2; dJ303F19.1; EGFL10; EGFL11; EGF-like protein 10; EGF-like protein 11; EGF-like-domain, multiple 10; EGF-like-domain, multiple 11; Epidermal growth factor-like protein 10; epidermal growth factor-like protein 11; eyes shut homolog (Drosophila); EYS; Protein eyes shut homolog; protein spacemaker homolog; RP25; SPAM; UNQ9424/PRO34591 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
346007 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T1H1 | |
EYS | |
Recombinant protein corresponding to Human EYS. Recombinant protein control fragment (Product #RP-94712). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction