Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ EZ Clip Wound Closure Kit
SDP

Catalog No. p-7163552 Shop All Stoelting Co Products
Click to view available options
Includes:
1000 Clips
200 Clips
Kit

Includes

Kit includes the Applier, case of 1,000 9mm Wound Clips and the Remover.

Stoelting™ EZ Clip Wound Closures Kit is ideal for rapidly closing wounds, providing maximum holding force with minimal trauma. Easier and faster to use than conventional sutures and needles.

Smaller teeth provide better clamping and wound closure of the animal's skin

  • Clips are easier to remove after healing
  • Less trauma to animals
  • Can be used with existing 9 mm appliers and removers

Use the Applier llike a staple gun to easily and quickly close the wound. When the wound is healed, the clips can be easily removed with the Clip Remover. All components may be purchased individually.

Racks of 9 mm clips are individually packaged in heat sealed, peelaway envelopes for cleanliness and ease of handling and storage. EZ Clips™ are packaged 20 on a rack, with 10 racks per box (200 total), or 50 racks per case (1000 total). The clips are pyrogen free and non-sterile. The autoclavable plastic packaging allows sterilization of the clips without removing them from the packaging.

The EZ Clip™ Kit includes the Applier, case of 1,000 9mm Wound Clips and the Remover.

Specifications

For Use With (Application) Neuroscience
Model Non-sterile
Includes Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.