Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ EZ-FPC™ Microorganism Preparations
SDP

Catalog No. 23003824 Shop All Microbiologics Products
Change view
Click to view available options
Species:
Alicyclobacillus acidoterrestris
Escherichia coli
Lactobacillus rhamnosus
ATCC®:
ATCC 43888
ATCC 49025
ATCC 9595
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species ATCC®
23-003-824 Alicyclobacillus acidoterrestris ATCC 49025
23-004-534 Escherichia coli ATCC 43888
23-003-826 Lactobacillus rhamnosus ATCC 9595
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-003-824 Supplier Microbiologics™ Supplier No. 0265FPC
This item is not returnable. View return policy

Includes

Product warranty and peel-off Certificate of Assay

Lyophilized microorganism preparations deliver a predetermined assay value for qualitative or quantitative daily process controls

  • Available in 100 to 1000CFU per preparation (qualitative) or 1000 to 10,000CFU per preparation (quantitative)
  • Daily process controls for food quality labs
  • Traceable to reference cultures

Order Info

Packaged in a vial of a single assayed microorganism (10 lyophilized pellets in each vial), with an attached Certificate of Assay

†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.

Specifications

ATCC® ATCC 49025
Format EZ-FPC
Product Type EZ-FPC
For Use With (Application) Lyophilized microorganism preparations deliver a predetermined assay value for qualitative or quantitative daily process controls.
Includes Product warranty and peel-off certificate of assay
Packaging Type Vial
Species Alicyclobacillus acidoterrestris
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.