Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EZ-Link™ Hydrazide-PEG4-Desthiobiotin, No-Weigh™ Format

Catalog No. pia39262
Click to view available options
Quantity:
5 x 1 mg

Thermo Scientific™ EZ-Link™ Hydrazide-PEG4-Desthiobiotin No-Weigh™ Format is a pegylated, carbonyl-reactive labeling reagent whose biotin-like group is elutable from streptavidin, enabling its use for cell surface glycoprotein labeling and purification.

  • Pegylated, carbonyl-reactive labeling reagent whose biotin-like group is elutable from streptavidin, enabling its use for cell surface glycoprotein labeling and purification.
  • Desthiobiotin - analog of biotin allows easy elution from streptavidin, an ideal feature for affinity purification applications
  • Glycoprotein labeling - tag glycosylated proteins at sialic acid residues to enable high recovery in pull-down assays with streptavidin beads
  • Cell surface labeling - tag and isolate cell surface glycoproteins; reagent does not permeate membranes of whole cells
  • Aldehyde-reactive - reacts with aldehydes formed by periodate-oxidation of sugar groups
  • Hydrazide-activated - perform reactions at pH 4 to 6 in buffers such as sodium acetate
  • Pegylated - spacer arm contains a hydrophilic, 4-unit, polyethylene glycol (PEG) group
  • Enhances solubility - pegylation imparts water solubility to the biotinylated molecule, helping to prevent aggregation of biotinylated antibodies stored in solution
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hydrazide-PEG4-Desthiobiotin
Quantity 5 x 1 mg
Content And Storage Store at 4°C.
Labeling Method Chemical Labeling
Shipping Condition Wet Ice
Product Line EZ-Link™
Cell Permeability Cell-Impermeant
Chemical Reactivity Carboxylic Acid, Ketone, Aldehyde
Reactive Moiety Amine, Hydrazide
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye Desthiobiotin
Spacer Extra-long, Pegylated
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.