Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EZ-Link™ Maleimide-PEG4-DBCO

Catalog No. FERC20041
Change view
Click to view available options
Quantity:
25 mg
10 x 1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERC20041 25 mg
FERC20044 10 x 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERC20041 Supplier Thermo Scientific™ Supplier No. C20041
Add to Cart
Edge
Add to Cart

A sulfhydryl reactive heterobifunctional crosslinker which contains a dibenzylcyclooctyne (DBCO) group for copper-free click chemistry applications. The thiol/sulfhdryl reactive maleimide can be used to label a variety of thiol containing moieties such as free cystines in proteins, peptides, or other (bio)molecules.

  • A sulfhydryl reactive heterobifunctional crosslinker which contains a dibenzylcyclooctyne (DBCO) group for copper-free click chemistry applications
  • The thiol/sulfhdryl reactive maleimide can be used to label a variety of thiol containing moieties such as free cystines in proteins, peptides, or other (bio)molecules
  • Once a protein or (bio)molecule is DBCO labeled, it can undergo a copper-free azide-alkyne cycloaddition reaction to form a stable triazole linkage with an azide labeled coupling partner
  • Can be used to create diverse conjugates with the biorthogonal coupling between an alkyne and azide (supplied separately)
  • The coupling or crosslinking procedure is typically done in two steps:
    • First, a protein or thiol containing molecule is labeled with a DBCO group and excess crosslinker is removed
    • Second, an azide containing moiety (prepared independently) is introduced into the same system as the DBCO moiety
    • Labeling occurs under biological conditions without the introduction of any additional reagents such as a copper catalyst
  • Reagent readily dissolves in water-miscible solvents for dilution into aqueous reaction mixtures
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Azide-Sulfhydryl
Cleavable No
Description Maleimide-PEG4-DBCO
Molecular Weight (g/mol) 647.74
PEGylated Yes
Spacer Arm Length 29.7 Å
Content And Storage Upon receipt store desiccated at -20°C.
Cell Permeability Not determined
Shipping Condition Wet Ice
Product Line EZ-Link No-Weigh
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Maleimide, DBCO
Spacer Medium (10 to 30 Å)
Form Solid
Quantity 25 mg
Solubility DMF, DMSO
Format Standard, Single-use
Water Soluble No
Product Type Crosslinker
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.