Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EZ-Link™ NHS-PEG12-Biotin, No-Weigh™ Format

Catalog No. p-4531565
Click to view available options
Quantity:
25 mg
10 x 1 mg
500 mg

Biotinylate protein amines and maximize solubility of proteins with this very long (56.0A) NHS-ester biotinylation reagent containing a pegylated, water-soluble spacer arm.

Thermo Scientific™ EZ-Link NHS-PEG12-Biotin is very long, pegylated biotinylation reagent for simple and efficient biotin labeling of antibodies, proteins and other primary amine-containing macromolecules.

Thermo Scientific No-Weigh products are specialty reagents provided in a pre-aliquoted format. The pre-weighed packaging helps prevent the loss of reagent reactivity and contamination over time by minimizing the repetitive opening and closing of the vial. The format enables use of a fresh vial of reagent each time, eliminating the hassle of weighing small amounts of reagents and reducing concerns over reagent stability.

  • Protein labeling: Biotinylate antibodies or other proteins for detection or purification using streptavidin probes or resins
  • Amine-reactive: Reacts with primary amines (-NH2), such as side-chain of lysines (K) or amino-termini of polypeptides
  • PEGylated: Spacer arm contains hydrophilic, 12-unit, polyethylene glycol (PEG) group
  • Enhances solubility: PEGylation imparts water solubility to biotinylated molecule, helping to prevent aggregation of biotinylated antibodies stored in solution
  • Irreversible: Forms permanent amide bonds; spacer arm cannot be cleaved
  • Very long reach: Spacer arm (total length added to target) is 56A0; this reduces steric hindrance when binding to avidin molecules
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 x 1 mg
Content And Storage Store desiccated at -5 to -30°C.
Shipping Condition Room Temperature
Product Line EZ-Link™, No-Weigh™
Solubility Water
Format Solid
Label Type Biotin & Analogs
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.