Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EZ-Link™ Sulfo-NHS-LC-Biotin
GSA_VA

Catalog No. PI21335 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mg
10 x 1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21335 100 mg
PIA39257 10 x 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PI21335 Supplier Thermo Scientific™ Supplier No. 21335
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ EZ-Link™ Sulfo-NHS-LC-Biotin is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.

Thermo Scientific™ EZ-Link Sulfo-NHS-LC-Biotin is an intermediate-length, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.

  • Protein labeling - biotinylate antibodies to facilitate immobilization, purification or detection using streptavidin resins or probes
  • Cell surface labeling - biotinylates only surface proteins of whole cells because the negatively charged reagent does not permeate cell membranes
  • Amine-reactive - reacts with primary amines (-NH2), such as lysine side-chains, or the amino-termini of polypeptides
  • Soluble - charged sulfo-NHS group increases reagent water solubility compared to ordinary NHS-ester compounds
  • Irreversible - forms permanent amide bonds; spacer arm cannot be cleaved
  • Medium length - spacer arm (total length added to target) is 22.4 angstroms; provides excellent balance between adequate length (minimal steric hindrance for biotin binding) and modest mass

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description EZ-Link Sulfo-NHS-LC-Biotin
Product Type Sulfo-NHS-LC-Biotin
Quantity 100 mg
Content And Storage Store desiccated at -20°C. Shipped at ambient temperature.
Product Line EZ-Link
Cell Permeability Cell-Impermeant
Chemical Reactivity Amine
Reactive Moiety Active Ester, Succinimidyl Ester, Sulfo-NHS Ester
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye Biotin
Spacer Mid-length
Label Type Biotin & Analogs
Show More Show Less
Can I make a Sulfo-NHS-LC-Biotin stock solution to use later?

No. NHS esters hydrolyze very quickly. The container must be brought to room temperature (RT) prior to opening, then the amount of biotinylation reagent weighed, dissolved and used immediately.

Can Sulfo-NHS-LC-Biotin be used in cell surface biotinylation?

Yes. Because this molecule dissolves readily in polar solutions and is itself charged by the sodium sulfoxide group on the succinimidyl ring, it cannot permeate the cell wall or membrane of an intact cell. As long as the cell is intact, only the primary amines on the surface will be biotinylated.

Why can't I perform biotinylation reactions in Tris buffer with Sulfo-NHS-LC-Biotin?

Tris is a primary amine, which will directly compete with protein primary amines for reaction with the NHS-ester activated biotin reagent. However, once a protein is labeled and desalted, Tris buffer can be used for storage or assay buffers, as appropriate for the protein and application.

What molar excess of Sulfo-NHS-LC-Biotin should I use to label antibodies and other proteins?

The molar excess to use depends on the concentration of the ligand to be biotinylated. Although the literature contains references that use between 2-50 molar excess of biotin, we recommend the following: 10-fold molar excess for 10 mg/mL solution of protein; 25-fold molar excess for 2 mg/mL solution of protein.

Can I dissolve the EZ-Link Sulfo-NHS-LC-Biotin, No-Weigh Format (Cat. No. A39257, 21335) in water for storage?

No, the EZ-Link Sulfo-NHS-LC-Biotin, No-Weigh Format (Cat. No. A39257) should not be dissolved in water for storage. We recommended storing EZ-Link Sulfo-NHS-LC-Biotin, No-Weigh Format at -20 degrees C with desiccant as it is very moisture sensitive. Furthermore, to avoid moisture condensation onto the product, equilibrate vial to room temperature before opening.

Can you provide the shelf-life for EZ-Link Sulfo-NHS-LC-Biotin?

EZ-Link Sulfo-NHS-LC-Biotin is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (-20 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

I received EZ-Link Sulfo-NHS-LC-Biotin (Cat. No. 21335) at ambient temperature, even though it is supposed to be stored at -20 degrees C. Is the performance of the product still reliable?

Yes, EZ-Link Sulfo-NHS-LC-Biotin is routinely shipped at room temperature. This product is stable at ambient temperatures for short periods of time.

What is the difference between EZ-Link Sulfo-NHS-LC-Biotin and Sulfo-NHS-LC-LC-Biotin?

EZ-Link Sulfo-NHS-LC-Biotin has a spacer arm length of 22.4 Angstroms while EZ-Link Sulfo-NHS-LC-LC-Biotin has a spacer arm length of 30.5 Angstroms.

Is there any advantage of choosing a Sulfo-NHS biotin with a longer spacer arm than EZ-Link Sulfo NHS Biotin?

Sometimes. The biotin has to insert 9 angstrom into the pocket of avidin, streptavidin or NeutrAvidin to bind. If steric hindrance may be a problem, using a biotinylation reagent with a longer spacer arm (Sulfo-NHS-LC Biotin or Sulfo-NHS-LC-LC Biotin) may result in greater purification or deteciton.

Why would I chose a Sulfo-NHS Biotin over its non-sulfonated counterpart?

The sodium sulfonate group lends water solubility to the reagent.
Note: The actual biotin moiety that is added is identical between NHS and Sulfo-NHS biotins that vary only in the addition of the sodium sulfonate on the leaving group. Researchers preferring a more water soluble biotin be added to their molecule should consider a PEGylated version of biotin.


WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.