Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EZ-Link™ Micro Sulfo-NHS-SS-Biotinylation Kit

Catalog No. pi21945
Click to view available options
Quantity:
8 reactions

Biotinylate protein amines with this cleavable, mid-length (24.3A), water-soluble, NHS-ester biotinylation reagent containing a cleavable disulfide bond in the spacer arm.

Thermo Scientific™ EZ-Link Sulfo-NHS-SS-Biotin enables simple and efficient biotin labeling of antibodies, proteins and any other primary amine-containing biomolecules in solution.

Highlights:

  • Cleavable analog of Sulfo-NHS-LC-Biotin
  • Can remove biotin label by cleaving spacer disulfide bond with reducing agents, such as DTT
  • Ideal for affinity purification experiments that require efficient elution from immobilized avidin supports (cleaving disulfide bond releases proteins)
  • Long spacer arm reduces steric hindrance for binding interactions
TRUSTED_SUSTAINABILITY

Specifications

Description EZ-Link Micro Sulfo-NHS-SS-Biotinylation Kit
Product Type Micro Sulfo-NHS-SS-Biotinylation Kit
Quantity 8 reactions
Content And Storage Sufficient For: 8 labeling reactions, each with 0.05–0.2 mg of antibody
• EZ-Link Sulfo-NHS-SS-Biotin, 8 x 1 mg
• PBS Pack (makes 500 mL), 1 pack
• Zeba Spin Desalting Columns (7K MWCO), 2 mL, 10 columns

Store biotin reagent at -20°C. Store remaining kit components at 4°C.
Labeling Scale 100 μg
Product Line EZ-Link™
Solubility Water
Label or Dye Biotin
Labeling Target Antibodies, Proteins
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.