Learn More
Thermo Scientific™ EZ-Link™ Sulfo-NHS-SS-Biotin, No Weigh Format

Description
Thermo Scientific™ EZ-Link Sulfo-NHS-SS-Biotin enables simple and efficient biotin labeling of antibodies, proteins and any other primary amine-containing biomolecules in solution.
Highlights:
- Cleavable analog of Sulfo-NHS-LC-Biotin
- Can remove biotin label by cleaving spacer disulfide bond with reducing agents, such as DTT
- Ideal for affinity purification experiments that require efficient elution from immobilized avidin supports (cleaving disulfide bond releases proteins)
- Long spacer arm reduces steric hindrance for binding interactions
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | EZ-Link Sulfo-NHS-SS-Biotin |
| Product Type | Sulfo-NHS-SS-Biotin |
| Quantity | 100 mg |
| Content And Storage | Store at 4°C. |
| Product Line | EZ-Link™ |
| Cell Permeability | Cell-Impermeant |
| Chemical Reactivity | Amine |
| Reactive Moiety | Active Ester, Succinimidyl Ester, Sulfo-NHS Ester |
| Solubility | DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water |
| Label or Dye | Biotin |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.