Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EZ-Link™ Sulfo-NHS-SS-Biotin, No Weigh Format

Catalog No. pia39258
Click to view available options
Quantity:
100 mg
10 x 1 mg

Thermo Scientific™ EZ-Link™ Sulfo-NHS-SS-Biotin No-Weigh™ Format is a water-soluble, NHS-ester biotinylation reagent whose spacer arm includes a cleavable disulfide bond for reversible labeling of proteins and cell surface primary amines.

Thermo Scientific™ EZ-Link Sulfo-NHS-SS-Biotin enables simple and efficient biotin labeling of antibodies, proteins and any other primary amine-containing biomolecules in solution.

  • Protein labeling - biotinylate antibodies to facilitate immobilization, purification or detection
  • Cell surface labeling - biotinylates only surface proteins of whole cells because the negatively charged reagent does not permeate cell membranes
  • Amine-reactive - reacts with primary amines (-NH2), such as lysine side-chains, or the amino-termini of polypeptides
  • Cleavable - disulfide bond in spacer arm allows the biotin label to be removed using reducing agents such as DTT; only a small sulfhydryl group remains attached to the molecule
  • Soluble - charged sulfo-NHS group increases reagent water solubility compared to ordinary NHS-ester compounds
  • Medium length - spacer arm (total length added to target) is 24.3 angstroms; it consists of the native biotin valeric acid group extended by a 7-atom chain
TRUSTED_SUSTAINABILITY

Specifications

Product Type Sulfo-NHS-SS-Biotin
Quantity 10 x 1 mg
Content And Storage Store at -20°C.
Labeling Method Chemical Labeling
Shipping Condition Wet Ice
Product Line EZ-Link™
Cell Permeability Cell-Impermeant
Chemical Reactivity Amine
Reactive Moiety Sulfo-NHS Ester
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye Biotin
Spacer Cleavable, 24.3 Å
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.