Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Innovating Science™ EZ Prep Chemicals (A-M)
Shop All Aldon Products

Click to view available options
Product Type:
Basic Fuschin 1%
Benedicts Reagent
Bromocresol Purple 0.4%
Bromothymol Blue 0.04%
Bromothymol Blue 0.1%
Bromothymol Blue 0.5%
Crystal Violet 1%
Eosin Y 1%
Hydrochloric Acid 1M
Malachite Green 1%
Methyl Green 1%
Methyl Orange 0.1%
Methylene Blue 0.1%
Methylene Blue 1%
Description
- Add to distilled water and stir to make the amount of solution indicated
- There is no need to handle chemicals as the chemicals are contained within the capsule
- Decrease preparation time since no weighing of chemicals is required
- Decrease disposal costs because no extra chemicals remain
Specifications
Specifications
Grade | 7 Lab |
Format | Capsule |
Product Type | Methyl Green 1% |
Quantity | 1 Capsule |
Color | White |
Languages | English |
For Use With (Application) | Preparations of chemical solutions |
Subject | Chemistry |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction