Learn More
Medtox™ Diagnostics EZ-SCREEN™ (HI) 9 Panel
Click to view available options
Detectable Analytes:
Oxazepam, Benzodiazepine Group
Description
- Screens for Amphetamine, Barbiturates, Benzodiazepine, Cocaine, Methamphetamine, Methadone, Opiates (2000), Oxycodone, Phencyclidine (PCP)
- Includes adulteration screen
- 510(k) cleared
- Quality management system conforms with ISO 13485
- Accurate screening results in minutes
- Made in the USA
Specifications
Specifications
CE Marker | No |
Clia Complexity | Moderate Complexity |
Detectable Analytes | Oxazepam, Benzodiazepine Group |
Disposable | Single-use |
For Use With (Application) | Drug Panel (Urine) |