Learn More
MilliporeSigma™ EZ-TFA (Universal Transcription Factor Assay, colorimetric)
Shop All MilliporeSigma ProductsDescription
Includes
- Transcription Factor Assay Plate
- Blocking Reagent
- 10X Annealing Buffer
- Substrate (TMB/E)
- Stop Solution
- TFA Buffer, 5X
- Gt X Rb, HRP Secondary Antibody
- Gt X Ms, HRP Secondary Antibody
Specifications
Specifications
| Content And Storage | Store components at the temperatures indicated on the labels. Various components to be stored at 4°C, -20°C, or -80°C |
| Detection Method | Chromogenic |
| For Use With (Application) | ELISA, Activity Assay |
| Quantity | 96 Assays |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.