Learn More
Restek™ EZ Twist Top Split/Splitless Injection Port Kits
SDP
Catalog No. 06611202 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Material:
Stainless Steel
Stainless Steel/Siltek™
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
06-611-202 Stainless Steel/Siltek™
06-611-201 Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-611-202 Supplier Restek™ Supplier No. 22178
Change Inlet Liners Faster and Easier, for Agilent 7890 GCs
  • Gas lines are attached to shell weldment bottom to help eliminate broken gas lines
  • Gas lines do not interfere with routine maintenance
  • Weldment removal tool allows for quick removal of the hot weldment without fingers ever touching hot surfaces
  • Weldment stays secure in the tool for reattachment
  • The shell weldment is designed for drop in installation
Specifications
Type EZ Twist Top Injection Ports
Material Stainless Steel/Siltek™
Quantity 1 Kit
For Use With (Equipment) Agilent 7890 GCs