Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ EZFluor™ 1-step Fluorescent Protein Gel Stain
Shop All MilliporeSigma Products
Click to view available options
Quantity:
1 L
4 L
Description
- Ready-to-use luminescent protein gel stain
- Requires only a single 5 to 30 minute staining step without fixation
- Destaining is optional
- Offers safer handling and ease of disposal, because it is an aqueous-based solution that does not contain hazardous methanol or acetic acid
- Passed environmental toxicity testing and is classified as non-hazardous to the environment under CCR Title 22 regulations
- Can detect as little as approximately 1 to 10 ng of protein per band depending on the staining method used
- Staining is fully compatible with mass spectrometry and Edman-based sequencing
- Absorbance: 288 nm
- Emission: 603 nm

Specifications
Specifications
Description | Absorbance: 288 nm; Emission: 603 nm |
Detection | Fluorescence |
Form | Liquid |
Quantity | 1 L |
Storage Requirements | Store at 2 to 8°C, protect from light |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction