Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EZH1 Monoclonal Antibody (3B6B9)
EZH1 Monoclonal Antibody (3B6B9)
Supplier: Thermo Scientific MA538518
Description
EZH1 Monoclonal Antibody for Western Blot, Flow, ELISA

Specifications
EZH1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q92800 | |
EZH1 | |
Purified recombinant fragment of human EZH1 (aa: 135-294) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
3B6B9 | |
Unconjugated | |
EZH1 | |
enhancer of zeste 1 polycomb repressive complex 2 subunit; enhancer of zeste homolog 1; enhancer of zeste homolog 1 (Drosophila); Enx2; ENX-2; Ezh1; Histone-lysine N-methyltransferase EZH1; hypothetical protein LOC664754; KIAA0388; KMT6B; si:ch211-250i3.1; zgc:136829 | |
Mouse | |
Protein G | |
RUO | |
2145 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction