Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EZH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115205
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115205 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115205 Supplier Invitrogen™ Supplier No. PA5115205
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total EZH1.

EZH1 is a component of a noncanonical Polycomb repressive complex-2 (PRC2) that mediates methylation of histone H3 lys27 (H3K27) and functions in the maintenance of embryonic stem cell pluripotency and plasticity (Shen et al., 2008 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen EZH1
Applications Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene EZH1
Gene Accession No. P70351, Q92800
Gene Alias enhancer of zeste 1 polycomb repressive complex 2 subunit; enhancer of zeste homolog 1; enhancer of zeste homolog 1 (Drosophila); Enx2; ENX-2; Ezh1; Histone-lysine N-methyltransferase EZH1; hypothetical protein LOC664754; KIAA0388; KMT6B; si:ch211-250i3.1; zgc:136829
Gene Symbols EZH1
Host Species Rabbit
Immunogen A synthesized peptide derived from human EZH1(Accession Q92800), corresponding to amino acid residues S172-G222.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14055, 2145
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.