Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EZH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518113
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518113 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518113 Supplier Invitrogen™ Supplier No. PA518113
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine and mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.

EZH1 is a component of a noncanonical Polycomb repressive complex-2 (PRC2) that mediates methylation of histone H3 lys27 (H3K27) and functions in the maintenance of embryonic stem cell pluripotency and plasticity (Shen et al., 2008 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen EZH1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene EZH1
Gene Accession No. Q92800
Gene Alias enhancer of zeste 1 polycomb repressive complex 2 subunit; enhancer of zeste homolog 1; enhancer of zeste homolog 1 (Drosophila); Enx2; ENX-2; Ezh1; Histone-lysine N-methyltransferase EZH1; hypothetical protein LOC664754; KIAA0388; KMT6B; si:ch211-250i3.1; zgc:136829
Gene Symbols EZH1
Host Species Goat
Immunogen Synthetic peptide sequence (EIPNPPTSKCITY) corresponding to the N-terminus amino acids of EZH1 (aa 2-15).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2145
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.