Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EZH1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$149.33 - $480.74
Specifications
| Antigen | EZH1 |
|---|---|
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
EZH1 Polyclonal specifically detects EZH1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| EZH1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EC 2.1.1.43, Enhancer of zeste homolog 1, enhancer of zeste homolog 1 (Drosophila), ENX-2, histone-lysine N-methyltransferase EZH1, KIAA0388enhancer of zeste (Drosophila) homolog 1, KMT6B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 160-280 of human EZH1 (NP_001982.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research | |
| PBS with 50% glycerol, pH7.3. | |
| 2145 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title