Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EZH2/KMT6 Antibody (10H8), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | EZH2/KMT6 |
---|---|
Clone | 10H8 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
EZH2/KMT6 Monoclonal antibody specifically detects EZH2/KMT6 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
EZH2/KMT6 | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 2.1.1, EC 2.1.1.43, enhancer of zeste (Drosophila) homolog 2, enhancer of zeste 2, Enhancer of zeste homolog 2, enhancer of zeste homolog 2 (Drosophila), ENX1, ENX-1KMT6EZH1KMT6A, histone-lysine N-methyltransferase EZH2, Lysine N-methyltransferase 6, MGC9169 | |
A synthesized peptide derived from Human EZH2/KMT6 [UniProt Q15910] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
10H8 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cancer, Chromatin Research, Phospho Specific, Prostate Cancer | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
2146 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title