Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EZH2 Monoclonal Antibody (AC22), eBioscience™, Invitrogen™

Catalog No. 14986737
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 mg
25 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-986-737 2 mg
14-986-780 25 μg
14-986-782 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14-986-737 Supplier Invitrogen™ Supplier No. 14986737
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The monoclonal antibody AC22 recognizes EZH2 (Enhancer of zeste homolog 2), a member of the polycomb group (PcG) proteins responsible for repression of gene expression through recognition and modification of histone methylation and chromatin structure. EZH2 comprises the catalytic subunit of the polycomb repressive complex 2 (PRC2) and directly represses gene expression through methylation of histone H3 on lysine 27 (H3K27) as well as direct protein targets such as GATA4. The PRC2 complex gene repression is necessary for proper development and differentiation. Underexpression of EZH2 is linked to abnormal developmental patterning and a loss of stem cell pluripotency, while overexpression of EZH2 is present in many tumors and correlates with poor prognosis in melanoma, breast, and prostate cancer. Applications Reported: This AC22 antibody has been reported for use in immunoprecipitation, western blotting, and immunocytochemistry. Applications Tested: This AC22 antibody has been tested by immunocytochemistry of methanol-fixed cells and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

EZH2 (Histone-lysine N-methyltransferase) is a member of the Polycomb-group (PcG) family. PcG family members form multimeric protein complexes, which are involved in maintaining the transcriptional repressive state of genes over successive cell generations. This protein associates with the embryonic ectoderm development protein, the VAV1 oncoprotein, and the X-linked nuclear protein. EZH2 may play a role in the hematopoietic and central nervous systems. Mutations affecting EZH2 can cause Weaver syndrome (WVS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen EZH2
Applications Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone AC22
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene EZH2
Gene Accession No. Q15910, Q61188
Gene Alias an; enhancer of zeste 2 polycomb repressive complex 2 subunit; enhancer of zeste homolog 2; enhancer of zeste homolog 2 (Drosophila); ENX 1; ENX1; Enx-1; Enx1h; EZH 2; EZH1; EZH2; EZH-2; EZH2b; histone-lysine N-methyltransferase EZH2; KMT6; KMT6A; lysine N-methyltransferase 6; mKIAA4065; OTTHUMP00000213774; OTTHUMP00000213778; OTTHUMP00000213779; OTTHUMP00000213780; WVS; WVS2
Gene Symbols EZH2
Host Species Mouse
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14056, 2146, 312299
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.