Learn More
EZH2 Mouse anti-Human, Mouse, Rat, Clone: 1F10A12, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | EZH2 |
---|---|
Clone | 1F10A12 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6899
![]() ![]() |
Proteintech Group Inc
664761IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody detected EZH2 protein as a single band with a molecular weight (MW) of 91-100 kDa in multiple cell lines. The phosphorylation may result in the higher molecular weight (calculated MW as 80-86 kDa).
FANCD2 (Fanconi anemia group D2 protein) is required for maintece of chromosomal stability. It promotes accurate and efficient pairing of homologs during meiosis. FANCD2 is involved in the repair of DNA double-stranded breaks, both by homologous recombination and single-stranded annealing. It is required for the targeting/stabilization of BLM to non-centromeric abnormal structures induced by replicative stress. FANCD2 promotes BRCA2/FANCD1 loading onto damaged chromatin. Defects in the FANCD2 gene result in Fanconi anemia complementation group D2. This disorder affects all bone marrow elements resulting in anemia, leukopenia, and thrombopenia.Specifications
EZH2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Enhancer of zeste homolog 2, ENX 1, EZH1, EZH2, KMT6, Lysine N methyltransferase 6 | |
Ezh2 | |
IgG1 | |
Protein G | |
Antibody |
1F10A12 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
Q15910, Q61188 | |
14056, 2146, 312299 | |
EZH2 Fusion Protein ag16789 | |
Primary | |
-20°C | |
EZH2 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.