Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EZH2 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501994134
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
50 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-4134 100 μL
50-199-4135 10 μL
50-199-4136 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-199-4134 Supplier Active Motif Supplier No. 39901
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EZH2 (Histone-lysine N-methyltransferase) is a member of the Polycomb-group (PcG) family. PcG family members form multimeric protein complexes, which are involved in maintaining the transcriptional repressive state of genes over successive cell generations. This protein associates with the embryonic ectoderm development protein, the VAV1 oncoprotein, and the X-linked nuclear protein. EZH2 may play a role in the hematopoietic and central nervous systems. Mutations affecting EZH2 can cause Weaver syndrome (WVS).

Specifications

Antigen EZH2
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Immunohistochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 30% glycerol and 0.035% sodium azide; pH
Gene EZH2
Gene Accession No. Q15910, Q61188
Gene Alias an; enhancer of zeste 2 polycomb repressive complex 2 subunit; Enhancer of zeste homolog 2; enhancer of zeste homolog 2 (Drosophila); ENX 1; ENX1; ENX-1; Enx1h; EZH 2; EZH1; Ezh2; EZH-2; EZH2b; Histone-lysine N-methyltransferase EZH2; KMT6; KMT6A; Lysine N-methyltransferase 6; mKIAA4065; OTTHUMP00000213774; OTTHUMP00000213778; OTTHUMP00000213779; OTTHUMP00000213780; WVS; WVS2
Gene Symbols EZH2
Host Species Rabbit
Immunogen This EZH2 antibody was raised against a recombinant fusion protein corresponding to amino acids 1-370 of mouse EZH2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14056, 2146
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.