Learn More
EZH2 Polyclonal Antibody, Active Motif

Rabbit Polyclonal Antibody
Supplier: Active Motif 65733

Description
Some products may be shipped at room temperature. This will not affect their stability or performance. Avoid repeated freeze/thaw cycles by aliquoting items into single-use fractions for storage at -20°C for up to 2 years. Keep all reagents on ice when not in storage.
EZH2 Polyclonal antibody specifically detects EZH2 in Human, Mouse samples. It is validated for Western BlotSpecifications
EZH2 | |
Polyclonal | |
whole serum with 30% glycerol and 0.035% sodium azide | |
Q15910, Q61188 | |
Rabbit | |
Affinity chromatography | |
RUO | |
14056, 2146 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
EZH2 | |
an; enhancer of zeste 2 polycomb repressive complex 2 subunit; enhancer of zeste homolog 2; enhancer of zeste homolog 2 (Drosophila); ENX 1; ENX1; Enx-1; Enx1h; EZH 2; EZH1; EZH2; EZH-2; EZH2b; histone-lysine N-methyltransferase EZH2; KMT6; KMT6A; lysine N-methyltransferase 6; mKIAA4065; OTTHUMP00000213774; OTTHUMP00000213778; OTTHUMP00000213779; OTTHUMP00000213780; WVS; WVS2 | |
This EZH2 antibody was raised against a recombinant fusion protein corresponding to amino acids 1-370 of mouse EZH2. | |
10 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.