Learn More
Invitrogen™ eZKine™ 4-Color Compensation Control Kit
Description
This eZKine™ 4-Color Compensation Control Kit is designed for compensation setup with eZKine™ Whole Blood Intracellular Cytokine Kits. It contains the necessary components to create single-color compensation controls for use with eZKine™ cocktails, including OneComp eBeads and 4 vials of antibody representing each of the 4 fluorochromes used in eZKine™ cocktails. OneComp eBeads can be stained with each of the 4 fluorochrome-conjuated antibodies to set up single-color compensation control tubes.
- Reactivity/Species: Human
- Reported Application: Flow Cytometric Analysis
Specifications
Specifications
| Format | Kit |
| No. of Tests | 25 Tests |
| Quantity | 25 tests |
| For Use With (Application) | Flow Cytometry |
| Product Line | eZKine |
| Product Type | Compensation Beads |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.