Learn More
Ezrin Antibody (2086C), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB72391V
Description
Ezrin Monoclonal antibody specifically detects Ezrin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, ImmunocytochemistrySpecifications
Ezrin | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry | |
2086C | |
Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
CVIL, CVL, Cytovillin, cytovillin 2, DKFZp762H157, EZR, ezrin, FLJ26216, p81, VIL2MGC1584, villin 2 (ezrin), villin-2 | |
E. coli-derived recombinant human Ezrin, Lys438-Arg562, Accession # P15311 | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Phospho Specific, Signal Transduction | |
7430 | |
Store at 4C in the dark. | |