Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ezrin Recombinant Rabbit Monoclonal Antibody (1L5O2)
GREENER_CHOICE

Catalog No. PIMA535229
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535229 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535229 Supplier Invitrogen™ Supplier No. MA535229
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: IEVQQMKAQA REEKHQKQLE RQQLETEKKR RETVEREKEQ MMREKEELML RLQDYEEKTK KAERELSEQI QRALQLEEER KRAQEEAERL EADRMAALRA K.

The cytoplasmic peripheral membrane protein encoded by this gene functions as a protein-tyrosine kinase substrate in microvilli. As a member of the ERM protein family, this protein serves as an intermediate between the plasma membrane and the actin cytoskeleton. This protein plays a key role in cell surface structure adhesion, migration and organization, and it has been implicated in various human cancers. A pseudogene located on chromosome 3 has been identified for this gene. Alternatively spliced variants have also been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ezrin
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 1L5O2
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ezr
Gene Accession No. P15311
Gene Alias AW146364; CVIL; CVL; Cytovillin; cytovillin 2; DKFZp762H157; epididymis secretory protein Li 105; ezr; ezr.L; ezr.S; EZRI; Ezrin; ezrin L homeolog; ezrin L homeolog; ezrin S homeolog; FLJ26216; HEL-S-105; MGC1584; p81; R75297; VIL2; villin 2; villin 2 (ezrin); villin-2; XELAEV_18027043mg
Gene Symbols Ezr
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7430
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.