Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ezrin, p Thr567 Antibody, Novus Biologicals™
SDP

Catalog No. p-7199719 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017836 0.025 mL
NB10081918 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017836 Supplier Novus Biologicals Supplier No. NB100819180.025ML
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ezrin Polyclonal specifically detects Ezrin in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Ezrin (p Thr567)
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide
Gene Accession No. P15311
Gene Alias CVIL, CVL, Cytovillin, cytovillin 2, DKFZp762H157, EZR, ezrin, FLJ26216, p81, VIL2MGC1584, villin 2 (ezrin), villin-2
Gene Symbols EZR
Host Species Rabbit
Immunogen The antiserum was produced against synthesized phosphopeptide derived from human Ezrin around the phosphorylation site of threonine 567 (Y-K-TP-L-R).
Molecular Weight of Antigen 81 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7430
Test Specificity Ezrin (specific to Phospho-Tyr567) detects endogenous levels of Ezrin only when phosphorylated at threonine 566.
Target Species Mouse, Rat, Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.