Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Ezrin Recombinant Rabbit Monoclonal Antibody (SAIC-37C-23)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547918
Description
Specificity: Recognises human EZR.
The cytoplasmic peripheral membrane protein encoded by this gene functions as a protein-tyrosine kinase substrate in microvilli. As a member of the ERM protein family, this protein serves as an intermediate between the plasma membrane and the actin cytoskeleton. This protein plays a key role in cell surface structure adhesion, migration and organization, and it has been implicated in various human cancers. A pseudogene located on chromosome 3 has been identified for this gene. Alternatively spliced variants have also been described for this gene.
Specifications
Ezrin | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
P15311 | |
Ezr | |
The KLH-conjugated peptide SGYLSSER derived from Ezrin, a protein likely involved in connections of major cytoskeletal structures to the plasma membrane. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG κ |
Flow Cytometry, Western Blot, Immunocytochemistry, Mass Spectrometry | |
SAIC-37C-23 | |
Unconjugated | |
Ezr | |
AW146364; CVIL; CVL; Cytovillin; cytovillin 2; DKFZp762H157; epididymis secretory protein Li 105; ezr; ezr.L; ezr.S; EZRI; Ezrin; ezrin L homeolog; ezrin L homeolog; ezrin S homeolog; FLJ26216; HEL-S-105; MGC1584; p81; R75297; VIL2; villin 2; villin 2 (ezrin); villin-2; XELAEV_18027043mg | |
Rabbit | |
Protein A | |
RUO | |
7430 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction