Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F-Box Only Protein 9Rabbit anti-Human, Polyclonal, Rockland™
SDP

Catalog No. 501991861
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1861 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-1861 Supplier Rockland Immunochemicals Supplier No. 600401918
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity-purified antibody is directed against human FBOX9 protein. A BLAST analysis was used to suggest cross reactivity with FBOX9 protein from human, dog, rat and chimpanzee based on 100% homology with the immunizing sequence. Expect partial reactivity with homologues from zebrafish, bovine and mouse (94% homology) as well as chicken (88%) and Xenopus laevis (82%). Reactivity is expected against all known isoforms of this protein, as the immunogen sequence is common to their carboxy-terminal ends. Reactivity against homologues from other sources is not known.

FBXO9 (F-box protein 9) is a Ubl conjugation pathway protein and it contains one F-box domain and one TPR repeat. FBXO9 may act as a potential sepsis biomarker or therapeutic target. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which functions in phosphorylation-dependent ubiquitination.

Specifications

Antigen F-Box Only Protein 9
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.53 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Fbxo9
Gene Accession No. Q9UK97
Gene Alias 9030401P18Rik; AA986398; Cross-immune reaction antigen 1; dJ341E18.2; F-box only protein 9; f-box protein 9; F-box protein Fbx9; FBX9; FBXO9; MNCb-2471; NY-REN-57; Renal carcinoma antigen NY-REN-57; VCIA1
Gene Symbols Fbxo9
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26268
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.