Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
F-box protein 15/FBXO15 Antibody (612834) [PE], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6035P
Description
F-box protein 15/FBXO15 Monoclonal antibody specifically detects F-box protein 15/FBXO15 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOFSpecifications
F-box protein 15/FBXO15 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
201456 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
612834 | |
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
F-box only protein 15, F-box protein 15, FBX15MGC39671 | |
E. coli-derived recombinant human F-box protein 15/FBXO15 isoform 2,, Asn298-Tyr434, Accession # Q8NCQ5-2 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction