Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant F. meningosepticum PNGase F Protein

Catalog No. p-7083341
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9109GH020 20 μg
9109GH100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 9109GH020 Supplier R&D Systems™ Supplier No. 9109GH020
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Effective removal of N-linked oligosaccharides from glycoproteins. Glycerol-free, suitable for downstream MS and HPLC.

Measured by its ability to deglycosylate ribonuclease B under denatured conditions. >50% ribonuclease B (10 μg) is deglycosylated by 2.5 ng rFmPNGase F within 30 minutes, as measured under the described conditions.

Specifications

Accession Number P21163
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Molecular Weight (g/mol) M.W. (Observed): 34 kDa, reducing conditions; M.W. (theoretical): 36 kDa
Quantity 20 μg
Source E. coli-derived f. meningosepticum PNGase F protein Ala41-Asn354 with N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Common Name PNGase F
Biological Activity Measured by its ability to deglycosylate ribonuclease B under denatured conditions. >50% ribonuclease B (10μg) is deglycosylated by 2.5 ng rFmPNGase F within 30 minutes, as measured under the described conditions.
Conjugate Unconjugated
Species F. meningosepticum
Recombinant Recombinant
Expression System E. coli-derived Ala41-Asn354 with N-terminal Met and 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PNGase F
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.