Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ p-Fluorobutyryl fentanyl (PFBF) Solution, 100 μg/mL in Methanol (), Sold by MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
0.5 mL
Description
A certified Snap-N-Spike™; solution suitable for use as starting material in calibrators and controls in para-Fluorobutyryl fentanyl LC/MS or GC/MS testing methods for clinical toxicology, urine drug testing or forensic analysis applications. para-Fluorobutyryl fentanyl, also known as PFBF and 4-FBF, is a designer drug and fentanyl analog that is abused recreationally.
Specifications
Specifications
| Packaging | Ampule |
| CAS | 67-56-1 |
| Concentration | 100 μg/mL in methanol |
| Linear Formula | C23H29FN2O |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Quantity | 0.5 mL |
| Formula Weight | 368.49 |
CERILLIANT is a registered trademark of Cerilliant Corporation
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction