Learn More
Thermo Scientific™ F1-ClipTip™ Multichannel Pipettes: PROMO



Description
Ergonomic Design
Remarkably Lightweight
Adjustable Comfort
Super Blow-out
Soft-touch Tip Ejection
Volume Lock Adjustment
Industry-leading Lifetime Warranty
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Compatible Tips | F1-ClipTip 200 |
Color | Yellow |
For Use With | F1-ClipTip™ 200 |
Model | F1-ClipTip™ |
Number of Channels | 12 |
Product Line | F1-ClipTip |
Type | Pipette |
Volume (Metric) | 10 to 100 μL |
Increments | 0.2 μL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.