Learn More
F10 Mouse anti-Human, Clone: OTI1D12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811723

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
F10 undergoes multiple processing steps before its preproprotein is converted to a mature two-chain form by the excision of the tripeptide RKR. Two chains of the factor are held together by 1 or more disulfide bonds; the light chain contains 2 EGF-like domains, while the heavy chain contains the catalytic domain which is structurally homologous to those of the other hemostatic serine proteases. The mature factor is activated by the cleavage of the activation peptide by factor IXa (in the intrisic pathway), or by factor VIIa (in the extrinsic pathway). The activated factor then converts prothrombin to thrombin in the presence of factor Va, Ca+2, and phospholipid during blood clotting. Mutations of this gene result in factor X deficiency, a hemorrhagic condition of variable severity.Specifications
F10 | |
Monoclonal | |
Unconjugated | |
F10 | |
FX, FXA | |
Mouse | |
Affinity Chromatography | |
RUO | |
2159 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1D12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P00742 | |
F10 | |
Human recombit protein fragment corresponding to amino acids 41-488 of human F10 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.