Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ F1142 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557441
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557441 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557441 Supplier Invitrogen™ Supplier No. PA557441
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SATVATVGQG ISNVIEKAET SLGIPSPSEI STEVKYVAGE TNAKENENSS PVAGAFGVFS TISTAVQSTG KSVISG Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.

FAM114A2 (Family With Sequence Similarity 114 Member A2) is a Protein Coding gene. Diseases associated with FAM114A2 include Postural Orthostatic Tachycardia Syndrome and Phencyclidine Abuse. Among its related pathways are Oncogenic MAPK signaling and HIV Life Cycle. Gene Ontology (GO) annotations related to this gene include purine nucleotide binding. An important paralog of this gene is FAM114A1. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen F1142
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM114A2
Gene Accession No. Q8VE88, Q9NRY5
Gene Alias 133K02; 1810073G14Rik; 9030624B09Rik; C5orf3; Fam114a2; family with sequence similarity 114 member A2; family with sequence similarity 114, member A2; protein FAM114A2; testis tissue sperm-binding protein Li 81P
Gene Symbols FAM114A2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human F1142. Recombinant protein control fragment (Product #RP-96806).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10827, 303155, 67726
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.